Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Food Coloring
SCI_ED

Catalog No. S02429 Shop All Aldon Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S02429 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02429 Supplier Innovating Science™ Supplier No. IS35039
Add to Cart
Add to Cart

Set of four food coloring in red, yellow, blue, and green.

Contains: Red #40, Yellow #5, Blue #1, and Green Dyes

Specifications

Grade 6+
Includes 50 mL Each Red, Yellow, Blue, Green Food Dye
Quantity 4 x 50 mL
Class Size 1 Class
Product Type Food Color Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.