Learn More
PIP™ Food Pro Bullet™ BioSoft™ Ear Plug
GREENER_CHOICE
Catalog No. 17467653 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Food Pro Bullet BioSoft Ear Plug; Corded: Corded
Catalog No.
17-467-653 Food Pro Bullet BioSoft Ear Plug; Corded: Corded
1 options
Catalog No. 17-467-653 Supplier PIP™ Supplier No. BSFD
  • $87.00
    $50.65 / Pack of 100
    Save $36.35
    42% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Metal detectable disposable corded foam ear plug made with BioSoft™ Technology.
  • PIP™ understands the importance of reducing waste to lower costs and minimize environmental impact
  • New, sustainable ECOSeries™ BioSoft™ ear plugs are made from bio-based technologies
  • BioSoft™ ear plug foam material provides the same fit and performance as conventional polyurethane or PVC materials, but with a lower carbon footprint
  • Innovative technology not only reduces emissions during manufacturing, but contains bio-based materials, making BioSoft™ more environmentally friendly at end of use
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class AL
Size Universal
High Visibility No
Quantity 1000 Plugs/Cs.
Noise Reduction Rating (NRR) 32 NRR
Corded Corded
Plug Shape Bullet
Disposable Single-use
Metal Detectable Yes
Dielectric No
EN 352.2002 EN 352-1
Includes Ear Plugs