Learn More
Dino Lite MS17TSW-F1 Foot Pedal for Capturing Images with DinoCapture
Supplier:  Dino Lite MS17TSWF1
Encompass_Preferred
MS17TSW-F1 is a shortcut command pedal that is convenient and efficient for capturing images with the DinoCapture software. The foot pedal is compatible with USB connected Dino-Lite and Dino-Eye units using either DinoXcope for Mac computers or DinoCapture 2.0 for PC, making it a single accessory for all users. The MS17TSW-F1 foot pedal is great for operators that require great attention on their work and may not have the ability to use their hands to capture an image. USB Cable Length: 9.5ftNote: MS17TSW-F1 will not function with Dino-Lite microscopes that are connected in Wireless modes. USB connection only.
Catalog No. NC1752756
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.