Learn More
Greenwich Safety For Responders QWIK-COVER™ Pre- and Post-Decon Dressing Kits
Maximize decontamination shower throughput by facilitating undress outside of the decon system
Supplier: Greenwich Safety DCN001LA
Description
Opaque decon garments fit over clothing, allowing ambulatory patients to disrobe in privacy. Perforations in these LLDPE plastic garments allow size adjustments in the overall length, arms, and head; as well as ensuring easy tear-away removal or conversion into a stretcher drape. Modesty flaps above the arm holes preserve privacy of the upper body. Color coded for easy identification of victim status. Packaged in easy-to-access zip-lock bags with pictographs for quick instructions.
Pre-Decon
- One pre-decon garment One large SECUR-ID™ garment bag One small SECUR-ID valuables bag
Post-Decon
- One post-decon garment 39 1/2 x 20 1/2 in. single-use Kimberly-Clark™ Dry-Up™ towel (amount varies for youth or large adult kits) Pair of spun bond polyethylene booties Molded nylon comb
Saddlebag Twin-Pack
- One pre-decon kit One post-decon kit Connected in an easy-to-carry saddlebag design
Specifications
| Dressing Kit | |
| Adult |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.