Learn More
Yamato Scientific America, Inc. Yamato 150L Forced Convection Oven, Programmable, 115V
Supplier:  Yamato Scientific America, Inc. DKN602C
Encompass_Preferred
Programmable 150L forced convection constant temperature oven with extended functions and safety features. Bestseller based on excellent performance and affordability. Superior temperature accuracy. Comes with observation window. Programmable PID controller for easy program settings (30-step, 3-pattern program controller with repeat functions). Fixed setting, Quick Auto stop, Auto stop, Auto start, Program operating modes with easy control capabilities. Overheating prevention and calibration off-set are possible with auxiliary functions. Easy to use and maintain. Equipped with built in exhaust ports. Operating temp ambient +10~260°C
Catalog No. 50-189-8004