Learn More
Eisco™ Force Board Kit
SCI_ED
Catalog No. S12944 Shop All Eisco Products
Change view
Click to view available options
Size: 40 x 55cm
Catalog No.
S12944 Size: 40 x 55cm
1 options
Catalog No. S12944 Supplier Eisco™ Supplier No. PH0345
Add to Cart
Add to Cart
Includes
ball-bearing pulleys, cotton reel, three hooks, mass set
Helps teacher demonstrate the principles of the parallelogram and polygon of forces.
  • Hard-wearing black board with printed grid
  • Detachable feet for storage
  • Bench standing
  • Compact and low maintenance
Specifications
Material Hard-wearing Blackboard
For Use With (Application) Demonstrate the principles of the parallelogram and polygon of forces
Product Type Force Board Kit
Includes detachable feet, ball-bearing pulleys, cotton reel, three hooks, mass set