Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Force Table
SDPSCI_ED

Catalog No. S00655 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S00655 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S00655 Supplier United Scientific™ Supplier No. FTE001
Add to Cart
Add to Cart
This item is not returnable. View return policy

Force Table provides the classroom with everything you need to teach about forces.

  • This student force table is a complete set.
  • Features a dual scale: an outer scale for accurate pulley positioning, and an inner scale for orienting a tracing paper overlay.
  • Set includes four complete weight sets (each with brass slotted weights, 2 x 5 g, 4 x 10 g, 4 x 20 g, and a 20 g brass weight hanger), four pulleys with clamps and two sets of strings that allows students to investigate the equilibrium of either three or four forces.
  • A 16 in. diameter lightweight, durable tabletop and sturdy cast iron support allow both ease of use and accuracy.
  • Includes activity guide.

Specifications

Grade 6 to 12
Format Kit
Product Type Force Table
Languages English
Subject Physics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.