Learn More
Vernier™ Force Table Adapter
SDPSCI_ED
Catalog No. S16153ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Application):
Educational use only
Catalog No. For Use With (Application)
S16153ND Educational use only
1 options
Catalog No. S16153ND Supplier Vernier™ Supplier No. FTADFS
Add to Cart
Add to Cart
Includes
Force Table Adapter
Use your Dual-Range Force Sensor with your force table for resolution of vectors experiments. Includes mounting clamp for tables up to 3/4 inch thick.
  • Use your Dual-Range Force Sensor with your force table for resolution of vectors experiments.
  • Includes mounting clamp for tables up to 3/4 inch thick.
Warranty and Services
5-year limited warranty
Specifications
Includes Mounting Clamp
Warranty 5 Year Limited
For Use With (Application) Educational use only
For Use With (Equipment) Dual-Range Force Sensor, Go Direct™ Force and Acceleration Sensor