Learn More
Memmert Memmert Universal Heating and Drying Laboratory Oven UF55. 55L - 230 Volt, Forced-Air Convection, Stainless Steel Construction with SingleDISPLAY digital controller.
Supplier: Memmert UF55 230V
Memmert Universal heating and drying Oven - this model is the 230 volt, forced-air convection, with SingleDISPLAY digital controller. Interior dimensions 400/400/291-W/H/D mm. Includes (1) Stainless Steel Grid Shelf, Quick Start Guide, Calibration Certificate and Power Cord - Working Range 10C above ambient to +300C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.