Learn More
Fine Science Tools Graefe Forceps, Stainless Steel, Angled, 10 cm
Supplier: Fine Science Tools 1104910
Tip Shape: Angled/ Tips: Serrated/ Tip Dimensions: 0.8 x 0.7mm/ Alloy / Material: Stainless Steel/ Length: 10cm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.