Learn More
Cytiva Forceps, Stainless Steel with Black Grips 1
Supplier:  Cytiva 51147
Encompass
For easy placement of membrane filters into filter funnels or petri dishes.•Flat surface•Avoid membrane damage•Stainless steelTips have a flat, smooth surface to avoid membrane filter damage. Stainless steel construction with polypropylene finger grips for a comfortable and secure hold. Autoclavable. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC0332089