Learn More
Med Vet International Miltex MeisterHand Mixter Thoracic Forceps, 11in, Right Angle Jaws, Longitudinal Serrations, German, MH25-814
Supplier:  Med Vet International MILMH25814
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Mixter Thoracic Forceps. Mixter Thoracic Forceps hold blood vessels for blunt dissection of the area that can be hard to reach by surgeons. They are made from premium-grade German stainless steel. Specifications: 11in, Right Angle Jaws, Longitudinal Serrations, MH25-814
Catalog No. 50-264-3489