Learn More
Med Vet International 3.5-in Economy Mosquito Forceps, Straight
Supplier:  Med Vet International 4500
Encompass_Preferred
Mosquito Forceps are used in multiple procedures as a hemostat for small blood vessels. The ratcheted finger ring handle provides a secure, locking grip to halt blood flow. They are not appropriate for use with large vessels or arteries.
Catalog No. NC2028707