Learn More
Med Vet International Miltex Wangensteen Tissue Forceps, 9in, Rounded Jaws W/Fin Cross Serrations, German, 16-80
Supplier:  Med Vet International MIL1680
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Wangensteen Tissue Forceps. Tissue Forceps can be used for fine handling of tissue and traction during dissection. They are made from premium-grade German stainless steel. Specifications: 9in, Rounded Jaws W/Fin Cross Serrations, 16-80
Catalog No. 50-252-2305