Learn More
United Scientific Supplies Forces and Simple Machines Kit
SDPSCI_ED
Catalog No. S96413 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Forces and Simple Machines Kit
Catalog No.
S96413 Forces and Simple Machines Kit
1 options
Catalog No. S96413 Supplier United Scientific Supplies Supplier No. FSMKIT01
Add to Cart
Add to Cart
Includes
Components include Force Board, Inclined Plane, Vector Board, an 11-piece weight set, spring scales, pulleys, levers, and a support base and rod. Also includes a comprehensive lab activities manual and a CD slide presentation of all experiments that can be performed with this set.
Contains a variety of custom-made and unique components.
Features accessories needed for 15 different activities.
Specifications
Product Type Kit, Forces and Simple Machines
Includes Components include force board, inclined plane, vector board, an 11-piece weight set, spring scales, pulleys, levers and a support base and rod. Also includes a comprehensive lab activities manual and a CD slide presentation of all experiments that can be performed with this set