Learn More
Description
Includes
Teacher's manual, student guide, analysis copy-masters, blood typing trays, four simulated blood samples (victim, suspect No. 1, suspect No. 2 and crime scene), ABO/Rh blood typing anti-sera, mixing sticks. Enough materials for 15 tests.
Blood type, an inherited characteristic, is valuable to know in that it affects medical procedures, such as surgery and transfusions, paternity testing, as well as serving as evidence in criminal investigations.
Uses Innovating Science Simulated Blood; safe for classroom use.
Specifications
Specifications
| Certifications/Compliance | 49 CFR 173.4 |
| Color | White |
| Grade | 7 to 14 |
| Age | 13+ years |
| For Use With (Application) | For forensic chemistry of blood typing |
| Includes | Blood typing trays, 4 simulated blood samples, crime scene, 1 set abo/rh blood typing anti-sera, 1 pkg mixing stick, Teacher's manual and student study guide copymasters |
| Quantity | 1 |
| Class Size | 15 Lab Groups |
| Product Type | Forensic Chemistry of Blood Types Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

