Learn More
Forensic Entomology Kit
SDPSCI_ED
Catalog No. S69005A Shop All Sirchie Finger Print Products
Change view
Click to view available options
Forensic entomology kit
Catalog No.
S69005A Forensic entomology kit
1 options
Catalog No. S69005A Supplier Sirchie Finger Print Supplier No. ENT1000
Add to Cart
Add to Cart
Use insects to help solve crimes.
Insects recovered from human remains can be a valuable tool for toxicological analysis and DNA of the victim.
  • Provides all of the tools, chemicals and containers needed to collect this valuable form of physical evidence
  • Insects and larvae retain traces of the fluids and blood of corpses after the soft tissues have decomposed
  • Recovered insects and larvae can reveal if a discovered body has been moved from the original crime scene, estimate a time of death, and recover valuable information about the victim and the possible crime
Specifications
For Use With (Application) For toxicological analysis and dna of the victim
Includes One butterfly net, fifteen 4 x 10 in. plastic specimen bags, evidence labels, plain labels, one 14 oz. kill jar, eight 2 oz. plastic jar, four 4 oz. plastic jar, one plastic trowel, one forceps - fine, straight, stainless, one forceps, featherweight, one
Product Type Forensic Entomology Kit