Learn More
Formaldehyde, 10% (v/v) (non-buffered), Histological Fixative, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Capacity | 4 L |
| Color | Colorless |
| Packaging Type | Natural Poly Bottle |
| Concentration | 10% (v/v) |
| For Use With (Equipment) | All Tissue Processors |
| Grade | Chemical Solution |
| Product Type | Formaldehyde |
| Solution Type | Formaldehyde |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.