Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formaldehyde, 10% (v/v), Buffered, pH 7.0, Histological Fixative, Ricca Chemical
SDP

Catalog No. 31905 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
4 L
10 L
20 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31905 20 L
31908 250 mL
R3190000500 500 mL
319032 1 L
31901 4 L
319025 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 31905 Supplier Ricca Chemical Company Supplier No. 31905
Add to Cart
Add to Cart

Specifications

Packaging Type Cubitainer
Concentration 10% (v/v)
For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Formaldehyde
Quantity 20 L
Solution Type Formaldehyde
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.