Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formaldehyde, 10% (v/v) Solution, Stabilized with approx. 3% Methanol, Histological Grade, Spectrum™ Chemical
SDP

Catalog No. 18610645 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
4 L
20 L
200 L
Packaging:
Drum
Poly Bottle
Poly Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-645 20 L Poly Drum
18-610-644 4 L Poly Bottle
18-610-646 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-610-645 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. HS00820LT
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50-00-0 , 7732-18-5 , 7558-79-4
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
IUPAC Name formaldehyde
SMILES C=O

Specifications

pH 4.0 to 7.0
Packaging Poly Drum
Quantity 20 L
Grade Histological
WARNING: Cancer - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.