Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formaldehyde, 37%, ACS Reagent (Formalin), Reagents

Catalog No. 01337954
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-954 20 L
01-337-956 10 L
01-337-955 1 L
01-337-953 4 L
01-337-952 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01-337-954 Supplier Reagents Holdings Llc Supplier No. C120850020F
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Formaldehyde, 37%, ACS Reagent (Formalin), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 50-00-0 , 67-56-1
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Formalin, Methanal
IUPAC Name water
SMILES O

Specifications

Color Colorless
Boiling Point 96°C
Packaging Cubitainer
Quantity 20 L
Specific Gravity 1.073
Grade ACS
Physical Form Liquid
Chemical Name or Material Formaldehyde, 37%, ACS Reagent (Formalin)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.