Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formaldehyde, 5% (v/v), Ricca Chemical
SDP

Catalog No. R31750004A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Poly natural
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R31750004A 4 L Poly natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R31750004A Supplier Ricca Chemical Company Supplier No. 31751
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 50-00-0 , 67-56-1
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
MDL Number MFCD00003274
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name water
SMILES O

Specifications

Boiling Point Approximately 100°C
Odor Pungent
Packaging Poly natural
Quantity 4 L
Solubility Information Solubility in Water: Infinite
Percent Purity 5% v/v
Grade Laboratory
Physical Form Liquid
Chemical Name or Material Formaldehyde, 5% (v/v) Aqueous Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.