Learn More
Abcam Formaldehyde Assay Kit, Fluorometric, Quantitative, 100 Tests
Description
- Detection method: Fluorescent
- Sensitivity: 1.5 μM
- Results type: Quantitative
- Sample types: Urine, Plasma, Serum, Other biological fluids, Cell Lysate
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Detection Location | In-Solution Detection |
| For Use With (Equipment) | Fluorometric Plate Reader |
| Format | Kit |
| No. of Reactions | 100 Tests |
| Target Molecule | Formaldehyde |
| Detection Method | Fluorescence |
| Sample Type | Urine, Plasma, Serum, Other biological fluids, Cell lysate |
| Quantity | 100 Tests |
| For Use With (Application) | Formaldehyde measurement |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.