Learn More
New Pig Formaldehyde-Neutralizing Spill Kit in Bucket
SDP
Catalog No. 18389127 Shop All New Pig Corporation Products
Change view
Click to view available options
Type: Formaldehyde-Neutralizing Spill Kit in Bucket
Catalog No.
18-389-127 Type: Formaldehyde-Neutralizing Spill Kit in Bucket
1 options
This item is not returnable. View return policy
Catalog No. 18-389-127 Supplier New Pig Supplier No. KIT501
  • $302.50
    $273.65 / Each of 1
    Save $28.85
    10% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Safely respond to formaldehyde spills with this specially equipped kit containing neutralizing absorbents and PPE.
  • PIG Formaldehyde Neutralizing Loose Absorbent quickly absorbs formaldehyde, formaldehyde-based fixatives, glutaraldehyde and other aldehyde solutions
  • Quickly solidifies liquid formaldehyde for easy cleanup and neutralizes the harmful vapors
  • Includes PPE and cleanup tools for safe, effective spill response
  • Screw-on, gasketed lid helps contain residual vapors after cleanup
  • Container with gasketed lid can be used for temporary storage of used absorbents
Specifications
Material Polyethylene