Learn More
Specifications
Specifications
| CAS Max % | 16.45, 78.37, 5.15, 0.03 |
| Decomposition Information | Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors |
| Color | Clear, colorless |
| pH | 8.0 to 9.0 |
| Packaging | Poly Bottle |
| Linear Formula | HCHO |
| Quantity | 500 mL |
| Formula Weight | 30.05 |
| Grade | Certified |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. May cause an allergic skin reaction. Causes serious eye damage. Harmful if inhaled. May cause cancer (inhalation). Causes damage to organs (respiratory system, skin). Harmful to aquatic life. |
| Precautionary Statement | GHS P Statement Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Avoid breathing gas, mist, vapors, spray. Wear protective clothing, protective gloves, eye protection, face protection. Wash exposed skin thoroughly after handling. Do not eat, drink or smoke when using this product. Use only outdoors or in a well-ventilated area. Contaminated work clothing must not be allowed out of the workplace. Avoid release to the environment. If swallowed: Rinse mouth. Do not induce vomiting. If on skin: Remove immediately all contaminated clothing. Rinse skin with water/shower. Immediately call a poison center/doctor. Wash contaminated clothing before reuse. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.