Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formaldehyde, Solution, ACS, 36.5% 38.0%, Spectrum™ Chemical
SDP

Catalog No. 18610653 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 mL
500 mL
1 L
4 L
20 L
200 L
Packaging:
Poly Drum
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-653 4 L Poly Drum
18-610-650 125 mL Poly Drum
18-610-651 500 mL Poly Drum
18-610-652 1 L Poly Drum
18-610-648 20 L Poly Drum
18-610-649 200 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-610-653 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F10804LTPL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50-00-0 , 7732-18-5 , 67-56-1

Specifications

Physical Form Liquid
CAS Max % 1
Packaging Poly Drum
Titratable Acid 0.006 meq/g
Color Colorless
Quantity 4 L
Percent Purity 36.5 to 38.0%
Grade ACS
Chemical Name or Material Formaldehyde, Solution
WARNING: Cancer - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.