Learn More
Formaldehyde, Unbuffered, Certified, 10% (v/v), LabChem™
Specifications
Specifications
| Chemical Name or Material | Formaldehyde |
| Name Note | Unbuffered |
| CAS | 7732-18-5, 50-00-0, 67-56-1 |
| CAS Max % | 95.05, 3.7, 1.25 |
| Decomposition Information | Carbon monoxide; Carbon dioxide |
| Color | Clear, colorless |
| Packaging | Cubitainer™ |
| Linear Formula | HCHO |
| Quantity | 20 L |
| Synonym | Formalin |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.