Learn More
Formaldehyde in Water, PT Proficiency Testing Material, MilliporeSigma™ Supelco™
Catalog No. 111002533 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
20 mL
Catalog No. Quantity
11-100-2533 20 mL
1 options
Catalog No. 11-100-2533 Supplier MilliporeSigma Supelco Supplier No. PE138020ML
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
This PT sample is supplied as a 20 mL concentrate. The solvent for this sample is water. Sample preparationTo prepare solution, partially fill a 1000-mL volumetric flask with ammonia-free, reagent-grade water (near 20 degrees C). Open ampule PEO-417 (be careful to avoid cuts on glass) and transfer 10.0 mL of concentrate to the volumetric flask. Fill the volumetric flask to volume with reagent-grade water and mix well. The sample is now ready for immediate analysis.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Ampule
Quantity 20 mL
Concentration or Composition (by Analyte or Components) Formaldehyde 5-500 mg/L