Learn More
Newcomer Supply Formalex Green Formalin Neutralizer, 5 gallon cube for drain disposal.
Supplier: Newcomer Supply ABFX05
Liquid formalin neutralizer that easily and safely neutralizes formalin waste to a non-hazardous, non-toxic solution for sink disposal. The improved formula eliminates any sludgy residue and does not require any pH adjustment. It contains no phosphates and is non-vapor forming. Use spray bottle for easy clean up of small countertop or bench spills!
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.