Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Formalin Dogfish Sharks
SDPSCI_ED

Catalog No. S37631 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Injection Type:
Double
Plain
Single
Triple
Length (English):
18 to 22 in.
22 to 27 in.
27+ in.
Type:
Preserved Specimen
Preserved Specimen, Pregnant
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Injection Type Length (English) Type
S37631 Single 27+ in. Preserved Specimen
S37630 Plain 27+ in. Preserved Specimen
S37632 Double 27+ in. Preserved Specimen
S37633 Triple 27+ in. Preserved Specimen
S72732 Double 27+ in. Preserved Specimen, Pregnant
S37634 Plain 22 to 27 in. Preserved Specimen
S37635 Single 22 to 27 in. Preserved Specimen
S37637 Triple 22 to 27 in. Preserved Specimen
S72731 Plain 18 to 22 in. Preserved Specimen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. S37631 Supplier Carolina™ Supplier No. 226721
Add to Cart
Add to Cart
This item is not returnable. View return policy

For use in exploring anatomy of cartilaginous fish. Carolina™ Formalin Dogfish Sharks are ideal for comparative and vertebrate anatomy classes.

  • Member of class Chondrichthyes
  • Mature specimens
  • Characteristics: cartilaginous skeleton, five pairs of gill slits, no swim bladder or operculum
  • Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid

Specifications

Type Preserved Specimen
Preservative Formalin
Injection Type Single
Length (English) 27+ in.
Length (Metric) 68.5+ cm
Quantity 1/Bag
Packaging Bag
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.