Learn More
Medchemexpress LLC Formamide deionized | 75-12-7 | 100.0% | 45.04 | CH3NO | 100mL
SDP
Supplier:  Medchemexpress LLC HYY0842B100ML
Encompass_Preferred
Formamide deionized (Methanamide deionizde) is an effective DNA denaturant that significantly reduces the stability of DNA in buffer solutions Formamide deionized can also be used as a decalcifying agent for rat cardiac cells Additionally Formamide deionized is widely used as a solvent or chemical raw material for ion compounds resins and plasticizers[1][2][3][4][5][6][7]
Catalog No. 50-002-67967
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.