Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formamide Solution NMR Reference Standard, 90 in DMSO-d6 (99.9 atom D), NMR tube size 3 mm, MilliporeSigma™ Supelco™

Catalog No. 111016598 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6598 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6598 Supplier MilliporeSigma Supelco Supplier No. 6133391EA

Specifications

CAS 75-12-7
Flash Point 87.8°C - closed cup
Linear Formula CH3 NO
Concentration 90% in DMSO- d 6 (99.9 atom% D)
MDL Number MFCD00007941
Type NMR Reference Standard
Quantity 1 Ea.
Formula Weight 45.04
Grade NMR Reference Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.