Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formate Standard, 1000 ppm HCOO-, Ricca Chemical
SDP

Catalog No. 321216 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
321216 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 321216 Supplier Ricca Chemical Company Supplier No. 321216
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64-18-6 , 7732-18-5
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O

Specifications

Packaging Natural Poly Bottle
CAS Min % 98, 12%
CAS Max % 100, 12%
Equivalence 1 mL = 1 mg HCOO
Physical Form Liquid
Color Colorless
pH Approximately 2 to 3
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Standard
Quantity 500 mL
Grade Laboratory
Chemical Name or Material Formate Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.