Learn More
Mopec™ Formaway Formalin Neutralizing Powder
SDP
Catalog No. 22445527 Shop All Mopec Products
Change view
Click to view available options
Weight (English):
14 lb.
38 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (English)
22-445-527 38 lb.
22-445-874 14 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-445-527 Supplier Mopec™ Supplier No. BG55032
Add to Cart
Add to Cart
Formaway Formalin Neutralizing Powder safely and easily neutralizes spent formalin waste to a non-hazardous and non-toxic solution in only 20 minutes. Contents are emptied directly into 1 gallon of spent 10% Formalin or 4% Glutaldehyde, reducing risk exposure to lab personnel.
This unique formula also eliminates drain-clogging polymers.
  • Collect waste in a designated collection container
  • Use a funnel with a 'screen insert' while collecting waste to remove tissue debris from waste
  • Add the entire contents of one Formaway bottle to each gallon of aldehyde waste
  • Secure lid on the collection container and agitate container to thoroughly mix solutions, make sure the agitation ultimately results in complete dissolution of Formaway Formalin Neutralizer Powder
  • Allow the mixed solution to stand for 20 to 25 minutes for the reaction to be complete
  • If required, test treated solution for any aldehyde residual prior to disposal (follow instructions for Formaway Formaldehyde Residual Screening Test Kit)
  • Pour treated waste into sanitary sewer and rinse container with cold tap water, preparing it for re-use
Specifications
Material Pre-treated Formalin Neutralizer
Type Formalin Neutralizer
Packaging Quantity 32 Bottles/Cs.
Weight (English) 38 lb.
Weight (Metric) 17.24 kg