Learn More
Molecular Research Center FORMAZOL 25 ML
SDP
Supplier:  Molecular Research Center FO 121
Encompass
FORMAzol,25 ml,specially purified and stabilized formamide for use as an RNA solubilizer,stable and can be used for RNA solubilization for two years when stored at 4degreeC or -20degreeC,RNA solubilized in FORMAzol is protected from degrada
Catalog No. NC9732950
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.