Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic Acid, 2% (w/v), Ricca Chemical
SDP

Catalog No. 32131 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
3213-1 4 L Natural Poly Bottle
3213-2-5 10 L Cubitainer
3213-5 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 3213-1 Supplier Ricca Chemical Company Supplier No. 32131
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) Mixture
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES OC=O

Specifications

CAS Min % 2.16, 96.0
Color Colorless
CAS Max % 2.25, 100.0
Concentration 2% (w/v)
Percent Purity 2% w/v
Grade Laboratory
Quantity 4 L
Packaging Natural Poly Bottle
Physical Form Liquid
Chemical Name or Material Formic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.