Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic Acid, 90.15%, MilliporeSigma™

Catalog No. 50947580 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-947-580 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-947-580 Supplier MilliporeSigma Supplier No. 1.10854.2500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Formic Acid for determination of viscosity acc. to DIN EN ISO 307.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.03
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym Methanoic acid solution, Formylic acid solution
IUPAC Name formic acid
SMILES OC=O

Specifications

Melting Point −9°C
Boiling Point 107°C (1013 hPa)
Density 1.2 g/cm3 (at 20°C)
Quantity 2.5 L
Explosion Limit 10 to 45.5% (V)
Flash Point 71°C
Ignition Point 485°C
Packaging Glass Bottle
Assay Percent Range 90.00 to 90.15%
Recommended Storage Store at 15 to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.