Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic Acid, 90%, Purified, Reagents

Catalog No. 01337596
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-596 500 mL
01-337-599 100 mL
01-337-598 2.5 L
01-337-597 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-337-596 Supplier Reagents Holdings Llc Supplier No. C2105000500A
Add to Cart
Add to Cart

Formic Acid, 90%, Purified, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-18-6 , 7732-18-5
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.03
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym Methanoic Acid, Formylic Acid, Aminic Acid
IUPAC Name formic acid
SMILES OC=O

Specifications

pH <2
Melting Point 8°C
Boiling Point 106°C
Color Colorless
Grade Reagent
Quantity 500 mL
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1.22
Chemical Name or Material Formic Acid, 90%, Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.