Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic Acid, 90%, Viscosity Grade, Reagents

Catalog No. 01337592
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-592 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-592 Supplier Reagents Holdings Llc Supplier No. C21050504A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Formic Acid, 90%, Viscosity Grade, Viscosity, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-18-6 , 7732-18-5
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.03
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym Methanoic Acid, Formylic Acid, Aminic Acid
IUPAC Name formic acid
SMILES OC=O

Specifications

pH 2.2
Melting Point 8°C
Boiling Point 106°C
Color Colorless
Quantity 4 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1.22
Chemical Name or Material Formic Acid, 90%, Viscosity Grade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.