Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic Acid 98 to 100%, For LC-MS LiChropur™, MilliporeSigma™

Catalog No. 01100264 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-100-264 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-100-264 Supplier MilliporeSigma Supplier No. 5.33002.0050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.03
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym Methanoic acid, Formylic acid
IUPAC Name formic acid
SMILES OC=O

Specifications

pH 2.2 (10 g/L water at 20°C)
Melting Point 4°C
Boiling Point 101°C at 1013 hPa
Density 1.22 g/cm3 at 20 °C
Quantity 50 mL
Explosion Limit 12 to 38% (V)
Flash Point 49.5 °C
Packaging Glass bottle
Vapor Pressure 42 hPa at 20 °C
Recommended Storage Store at 15 to 25 °C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.