Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic Acid, ACS, 88%, Spectrum™ Chemical
SDP

Catalog No. 18610661 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Drum
Poly-Coated Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-661 4 L Poly-Coated Glass Bottle
18-610-663 100 mL Amber Glass Bottle
18-610-666 500 mL Amber Glass Bottle
18-610-664 2.5 L Glass Bottle
18-610-662 20 L Poly Drum
18-610-665 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-610-661 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F10904LTPV
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64-18-6 , 7732-18-5

Specifications

CAS Max % 1
Percent Purity 88%
Grade ACS
Quantity 4 L
Packaging Poly-Coated Glass Bottle
Residue after Evaporation 2e-05

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.