Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic Acid (Aldehyde-Free/Sequencing), Fisher BioReagents™

Catalog No. bp1215500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP1215-500 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP1215-500
Formic Acid is used in Maxam-Gilbert DNA sequencing reactions and other biological applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-18-6 , 7732-18-5
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O

Specifications

pH 2.1
Melting Point 8°C
Boiling Point 101°C
Color Colorless
Assay Percent Range ≥88.0 %
Linear Formula HCOOH
UN Number 1779
Merck Index 15, 4269
Formula Weight 46.02
Percent Purity ≥88.0%
Grade Sequencing
Density 1.220g/cm³
Quantity 500 mL
Packaging Poly Bottle
Physical Form Liquid
Residue after Evaporation 0.002% max.
Vapor Pressure 44mbar at 20°C
Chemical Name or Material Formic Acid, Aldehyde-free
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Use only under a chemical fume hood. Use only non-sparking tools. Use explosion-proof equipment. Do not breathe vapors/dust. Do not ingest. Take precautionary measures against static discharges. Wear personal protective equipment. Do not get in eyes, on skin, or on clothing. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes.

NFPA

Health:3
Flammability:2
Instability:0
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.