Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic acid-d2, for NMR, 99+ atom % D, 95% solution in D2O

Catalog No. AC233400010
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC233400010 1 g Glass Bottle
AC233400100 10 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC233400010 Supplier Thermo Scientific Chemicals Supplier No. 233400010
Get 1 Extra Chemical When You Buy 2
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 920-42-3 | CH2O2 | 48.037 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 920-42-3
Molecular Formula CH2O2
Molecular Weight (g/mol) 48.037
MDL Number MFCD00037363
InChI Key BDAGIHXWWSANSR-PFUFQJKNSA-N
Synonym formic acid-d2, 2h formic 2 acid, dcood, 2h2 formic acid, formic-d acid-d 6ci,7ci,8ci,9ci, formic-d acid-d, 95 wt% in deuterium oxide, formic acid-d2, 95 wt. % in d2o, 98 atom % d
PubChem CID 123092
IUPAC Name deuterio deuterioformate
SMILES C(=O)O

Specifications

Physical Form Liquid
Color Colorless
Packaging Glass Bottle
Quantity 1 g
Flash Point 50°C
Specific Gravity 1.27
Density 1.2700g/mL
Deuterium (D) 99.0% min.
Melting Point 5.9°C
Boiling Point 101.0°C
Linear Formula DCOOD
Solubility Information Solubility in water: miscible.
Formula Weight 48.04
Percent Purity ≥94.0%
Grade NMR
Chemical Name or Material Formic acid-d2, 99+ atom % D (95% in D2O)
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTRE or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-057-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.