Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic acid, Eluent additive for LC-MS, Solstice

Catalog No. 60048227 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
10 x 1 mL
1 L
Packaging:
Ampoule
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-227 10 x 1 mL Ampoule
60-047-107 50 mL Glass Bottle
60-006-14 1 L Glass Bottle
60-006-15 50 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-048-227 Supplier Solstice Supplier No. 5630210X1ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eluent additive for LC-MS

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
MDL Number MFCD00003297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O

Specifications

Melting Point 8.2°C to 8.4°C (literature)
Boiling Point 100.4°C
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Density 1.22 g/mL at 25 °C(lit.)
Quantity 10 x 1 mL
Explosion Limit 0.57
Flash Point 49.5°C
Ignition Point 1004°F
Packaging Ampoule
Linear Formula H2O
Vapor Density 1.6 (vs air)
UN Number UN1779
Vapor Pressure 44.8 mmHg (20°C)
Beilstein 1209246
Formula Weight 46.03
Percent Purity >97.5%
Grade LC-MS
Chemical Name or Material Formic acid, Eluent additive for LC-MS
Show More Show Less
Hazard Statement P210-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P403 + P233
ShelfLife 1080 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Formic acid
EINECSNumber 200-579-1
RTECSNumber 200-579-1
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.