Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formic Acid, LC-MS Ultra, Solstice

Catalog No. 60044843 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2 mL
Packaging:
Ampoule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-843 2 mL Ampoule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-044-843 Supplier Solstice Supplier No. 142652ML
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
MDL Number MFCD00003297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O

Specifications

Melting Point 4°C
Boiling Point 100.4°C
Color Colorless
Density 1.22g/mL at 25°C
Quantity 2 mL
Explosion Limit 0.57
Flash Point 49°C (120.2°F)
Ignition Point 1004°F
Packaging Ampoule
Physical Form Liquid
Refractive Index n20/D 1.370(lit.)
Linear Formula HCOOH
Vapor Density 1.6 (vs air)
UN Number UN1779
Vapor Pressure 44.8mmHg (20°C)
Beilstein 1209246
Formula Weight 46.03g/mol
Percent Purity ∼98% (T)
Grade LC-MS Ultra
Chemical Name or Material Formic Acid
Show More Show Less
Hazard Statement P210-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P403 + P233
Precautionary Statement H226-H302-H314-H331
DOTInformation 3(8)/PGII
EINECSNumber 200-579-1
RTECSNumber LQ4900000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.