Learn More
Selleck Chemical LLC Formoterol Hemifumarate 10mg 43229-80-7 Eformoterol, CGP 25827A, NSC 299587, YM 08316
SDP
Supplier:  Selleck Chemical LLC S202010MG
Encompass_Preferred
Formoterol Hemifumarate (Eformoterol, CGP 25827A, NSC 299587, YM 08316) is a potent, selective and long-acting ?2-adrenoceptor agonist used in the management of asthma and chronic obstructive pulmonary disease(COPD). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1886
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.