Learn More
Cayman Chemical Forskolin, 66575-29-9, 5 mg
SDP
Supplier:  Cayman Chemical 110185
Encompass_Preferred
A naturally occurring diterpene that directly activates adenylyl cyclase to raise levels of cAMP in a wide variety of cell types; binds to type 1 adenylyl cyclase membranes with an IC50 value of 41 nM and demonstrates an EC50 value of 0.5 μM in an activation assay
Catalog No. NC0302886
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.