Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Forskolin, 98+%, 50 mg, Thermo Scientific Chemicals PROMO

Catalog No. J63292MFPM
Change view
Click to view available options
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
J63292MFPM 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. J63292MFPM Supplier Thermo Scientific Chemicals Supplier No. J63292MFPR

Forskolin, CAS # 66575-29-9, is a diterpenoid compound isolated from Indian Coleus that acts as a strong activator of the adenylate cyclase system.

General Description

  • Forskolin is a diterpenoid and cyclic ketone, found naturally as a plant metabolite
  • It is a protein kinase A agonist and a cyclic ketone
  • Forskolin is an antihypertensive and muscle relaxant, it can also promote the release of pituitary hormones and prevent platelet aggregation

Application

  • Forskolin assists in neuron differentiation in vitro
  • Useful for studies of cell metabolism and development, there is some evidence of its usefulness in the study of glaucoma
  • It is intended for cell culture and biochemistry applications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66575-29-9
Molecular Formula C22H34O7
Molecular Weight (g/mol) 410.507 g/mol
MDL Number MFCD00082317
InChI Key OHCQJHSOBUTRHG-KGGHGJDLSA-N
Synonym forskolin, colforsin, coleonol, colforsina, colforsine, colforsinum, boforsin, colforsine french, colforsinum latin, colforsina spanish
PubChem CID 47936
ChEBI CHEBI:42471
IUPAC Name [(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate
SMILES CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O

Specifications

Color White
Quantity 50 mg
Beilstein 1692716
Merck Index 14,2476
Solubility Information Soluble in ethanol,dimethylsulfoxide and chloroform; Insolule in Water.
Formula Weight 410.51 g/mol
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material Forskolin
Hazard Category H312
Hazard Statement GHS H Statement: H315-H319-H335. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement P280-P302+P352-P312-P363-P501c
EINECSNumber 266-410-9
RTECSNumber QL6150000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.