CAS RN 66575-29-9
MilliporeSigma™ Calbiochem™ Forskolin, Coleus forskohlii
Forskolin, CAS 66575-29-9, is a cell-permeable diterpene inotropic agent that acts as an activator of adenylate cyclase (EC50 = 4 mM).
Forskolin, (from Coleus forskohlii), MP Biomedicals™
CAS: 66575-29-9 Molecular Formula: C22H34O7 Molecular Weight (g/mol): 410.507 MDL Number: MFCD00082317 InChI Key: OHCQJHSOBUTRHG-KGGHGJDLSA-N Synonym: forskolin,colforsin,coleonol,colforsina,colforsine,colforsinum,boforsin,colforsine french,colforsinum latin,colforsina spanish PubChem CID: 47936 ChEBI: CHEBI:42471 IUPAC Name: [(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate SMILES: CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O
Forskolin, 98+%
CAS: 66575-29-9 Molecular Formula: C22H34O7 Molecular Weight (g/mol): 410.507 MDL Number: MFCD00082317 InChI Key: OHCQJHSOBUTRHG-KGGHGJDLSA-N Synonym: forskolin,colforsin,coleonol,colforsina,colforsine,colforsinum,boforsin,colforsine french,colforsinum latin,colforsina spanish PubChem CID: 47936 ChEBI: CHEBI:42471 IUPAC Name: [(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate SMILES: CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O
Forskolin 95.0+%, TCI America™
CAS: 66575-29-9 Molecular Formula: C22H34O7 Molecular Weight (g/mol): 410.507 MDL Number: MFCD00082317 InChI Key: OHCQJHSOBUTRHG-KGGHGJDLSA-N Synonym: forskolin,colforsin,coleonol,colforsina,colforsine,colforsinum,boforsin,colforsine french,colforsinum latin,colforsina spanish PubChem CID: 47936 ChEBI: CHEBI:42471 IUPAC Name: [(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate SMILES: CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O
R&D Systems™ Forskolin
A Tocris product, part of R&D Systems. Forskolin synthesized to Ancillary Material Grade
R&D Systems™ Forskolin
CAS: 66575-29-9 Molecular Formula: C22H34O7
Forskolin (From Coleus Forskohlii), Fisher BioReagents
CAS: 66575-29-9 Molecular Formula: C22H34O7 Molecular Weight (g/mol): 410.507 InChI Key: OHCQJHSOBUTRHG-KGGHGJDLSA-N Synonym: forskolin,colforsin,coleonol,colforsina,colforsine,colforsinum,boforsin,colforsine french,colforsinum latin,colforsina spanish PubChem CID: 47936 ChEBI: CHEBI:42471 IUPAC Name: [(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate SMILES: CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O
Forskolin, 98+%, 10 mg, Thermo Scientific Chemicals PROMO
CAS: 66575-29-9 Molecular Formula: C22H34O7 Molecular Weight (g/mol): 410.507 g/mol MDL Number: MFCD00082317 InChI Key: OHCQJHSOBUTRHG-KGGHGJDLSA-N Synonym: forskolin,colforsin,coleonol,colforsina,colforsine,colforsinum,boforsin,colforsine french,colforsinum latin,colforsina spanish PubChem CID: 47936 ChEBI: CHEBI:42471 IUPAC Name: [(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate SMILES: CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O
Forskolin, 98+%, 50 mg, Thermo Scientific Chemicals PROMO
CAS: 66575-29-9 Molecular Formula: C22H34O7 Molecular Weight (g/mol): 410.507 g/mol MDL Number: MFCD00082317 InChI Key: OHCQJHSOBUTRHG-KGGHGJDLSA-N Synonym: forskolin,colforsin,coleonol,colforsina,colforsine,colforsinum,boforsin,colforsine french,colforsinum latin,colforsina spanish PubChem CID: 47936 ChEBI: CHEBI:42471 IUPAC Name: [(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate SMILES: CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O