Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Forskolin (From Coleus Forskohlii), Fisher BioReagents

Catalog No. BP25201 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Packaging:
Amber Glass
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP25201 1 mg Amber Glass
BP25205 5 mg Amber Glass
BP252010 10 mg Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP25201
Forskolin is a molecular biology reagent. Forskolin functions as an antihypertensive and vasodilator. Adenylcyclase activator.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66575-29-9
Molecular Formula C22H34O7
Molecular Weight (g/mol) 410.507
InChI Key OHCQJHSOBUTRHG-KGGHGJDLSA-N
Synonym forskolin, colforsin, coleonol, colforsina, colforsine, colforsinum, boforsin, colforsine french, colforsinum latin, colforsina spanish
PubChem CID 47936
ChEBI CHEBI:42471
IUPAC Name [(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate
SMILES CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O

Specifications

Melting Point 228°C
Color White
Packaging Amber Glass
Quantity 1 mg
Merck Index 15, 2460
Formula Weight 410.5
Physical Form Solid
Chemical Name or Material Forskolin, From Coleus forskohlii
Safety handling Check Alert storage code Red
Recommended Storage RT, desiccate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.